missing translation for 'onlineSavingsMsg'
Learn More

Human CD300f/LMIR3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7061332
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18727047 25 μg 25µg
18002332 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18727047 Supplier R&D Systems Supplier No. AF2774SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

CD300f/LMIR3/CD300LF Polyclonal specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300f/LMIR3/CD300LF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TDQ1
Gene Alias CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3
Gene Symbols CD300LF
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD300f/LMIR3 Tyr16-Leu155 (Val19Ala) Accession # Q8TDQ1
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Test Specificity Detects human CD300f/LMIR3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) LMIR5 and rhLMIR4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.