missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD30/TNFRSF8 DuoSet ELISA

Product Code. 30000456
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 30000456

Brand: R&D Systems™ DY612605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

For the development of sandwich ELISAs to measure natural and recombinant human CD30

The Human CD30/TNFRSF8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CD30/TNFRSF8.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 93.80 to 6000 pg/mL
Detectable Analytes CD30/TNFRSF8
Assay Solid Phase Sandwich ELISA
Conjugate Biotin
Cross Reactivity <50%cross-species reactivity observed with species tested
Immunoassay Kit Format 96-well Strip Plate
Product Type ELISA Development Kit
Sample Type Cell Culture Supernatant
Sufficient For To run ELISAs on at least 5 x 96-well plates
For Use With (Application) ELISA
Gene ID (Entrez) 21941, 943, 25069
Host Species Human
Kit Contents Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Quantity 5 Plates
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.