missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD30 Ligand / TNFSF8 Protein, Mouse IgG2a Fc Tag, low endotoxin

Product Code. 16467389
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467389 1 mg 1mg
16457389 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16467389 Supplier ACROBiosystems Supplier No. CDLH525b1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

low endotoxin

CD30 ligand (CD30L) and CD30 are interacting cellsurface glycoproteins that are members of the tumour necrosis factor (TNF) and tumour necrosis factor receptor (TNFR) superfamilies. In humans, CD30L is expressed at high levels on activated T cells. There is conflicting evidence with regard to cell surface expression of CD30L on primary human B cells. An initial report of constitutive CD30L expression on peripheral blood B cells33 has not been substantiated.

This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_001235.1
Molecular Weight (g/mol) 46.5 kDa
Quantity 1 mg
Source Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H525b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (Accession # P32971-1).
Immunogen Gln 63 - Asp 234
Storage Requirements -20°C
Gene Alias CD30-L,CD153,TNFSF8,CD30L,CD30LG,CD153 antigen,CD30 antigen ligand,CD30 Ligand
Common Name CD30 ligand
Conjugate Unconjugated
Species Human
Protein Tag Mouse IgG Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 0.1 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.