missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD3 epsilon F(ab')2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£195.00 - £1785.00
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Clone | UCHT1R-FAB2-TEV/His |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18740447
![]() |
R&D Systems
MAB100R-FAB2-SP |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792547
![]() |
R&D Systems
MAB100R-FAB2-100 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764247
![]() |
R&D Systems
MAB100R-FAB2-500 |
500 μg |
£1785.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762097
![]() |
R&D Systems
MAB100R-FAB2-01M |
1 mg |
£1685.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Human samples. It is validated for Flow CytometrySpecifications
| CD3 epsilon | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Human thymocytes | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| UCHT1R-FAB2-TEV/His | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 916 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title