missing translation for 'onlineSavingsMsg'
Learn More

Human CD3 epsilon Antibody, R&D Systems™
GREENER_CHOICE

Recombinant Monoclonal Antibody

£203.00 - £1845.00

Specifications

Antigen CD3
Clone UCHT1R
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, T Cell Stimulation, Immunocytochemistry 1-25 ug/mL
Applications Western Blot, Flow Cytometry, T-Cell Activation, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752729
GREENER_CHOICE
View Documents
R&D Systems
MAB100R-SP
25 μg
£203.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18732937
GREENER_CHOICE
View Documents
R&D Systems
MAB100R-100
100 μg
£452.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18100321
View Documents
R&D Systems
MAB100R-500
500 ug
£1845.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18112411
View Documents
R&D Systems
MAB100R-01M
1 mg
£1680.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Western Blot, Flow Cytometry, T Cell Stimulation, Immunocytochemistry.
Specifications

Specifications

CD3
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, T Cell Stimulation, Immunocytochemistry 1-25 ug/mL
Monoclonal
Purified
RUO
CD_antigen: CD3e, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, IMD18, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
CD3E
IgG1
Protein A or G purified from cell culture supernatant
Recognizes the epsilon-chain of the CD3/T cell antigen receptor complex (McMichael, A.J. et al. (1987) Leucocyte Typing III: White Cell Differentiation Antigens, Oxford University Press, New York; Knapp, W. et al. (1989) Leucocyte Typing IV: White Cell Differentiation Antigens, Oxford University Press, New York; Schlossman, S. et al. (1995) Leucocyte Typing V: White Cell Differentiation Antigens, Oxford University Press, New York).
UCHT1R
Western Blot, Flow Cytometry, T-Cell Activation, Immunocytochemistry
Unconjugated
Mouse
Human
916
Human thymocytes
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.