Learn More
Gibco™ Human CD25 (IL-2R alpha) (soluble) Recombinant Protein, PeproTech®
Description
200-02RC-1MG will be provided as 2 x 500 μg (200-02RC-500UG). Recombinant Human sIL-2 Receptor alpha is a 24.8 kDa protein containing 219 amino acid residues consisting of only the extracellular domain of IL-2Ralpha. As a result of glycosylation, Recombinant Human sIL-2 Receptor alpha migrates with an apparent molecular mass of approximately 40-50 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P01589 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3559 |
| Molecular Weight (g/mol) | 24.8 kDa |
| Name | Human CD25 (IL-2 R alpha) (soluble) |
| Quantity | 100 μg |
| Source | CHO cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.