missing translation for 'onlineSavingsMsg'
Learn More

Human CD25/IL-2R alpha PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 6 publications

£174.00 - £281.00

Specifications

Antigen CD25/IL-2R alpha
Clone 24212
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752649
GREENER_CHOICE
View Documents
R&D Systems
FAB1020P-025
25 Tests
£174.00
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18739336
GREENER_CHOICE
View Documents
R&D Systems
FAB1020P-100
100 Tests
£281.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD25/IL-2R alpha
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
IL2RA
IgG2a
Protein A or G purified from hybridoma culture supernatant
Detects human CD25/IL-2 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R gamma or rhIL-15 R alpha is observed.
24212
Flow Cytometry
R-PE
Mouse
Human
P01589
3559
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha Glu22-Cys213 Accession # P01589
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.