missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CD25/IL-2 R alpha QuicKit ELISA
Specifications
Specifications
| Specificity | Natural and recombinant human IL-2 R alpha |
| Target Species | Human |
| Gene ID (Entrez) | 3559 |
| Quantity | 1 Pack (6 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?