missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human CD23,FCER2 ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Description
- Recognizes human CD23
- CD23 (low affinity immunoglobulin ε Fc receptor) is a transmembrane glycoprotein present on subpopulation of B lymphocytes in germinal centers, EBV-transformed B-lymphoblastoid cell lines, follicular dendritic cells and a subpopulation of peripheral blood cells
- Essential roles in regulation of IgE production and differentiation of B-cells
- Can be cleaved into soluble fragments (sCD23) of various sizes displaying pleiotropic biological activities
- Increased plasma levels of sCD23 reported in allergic disorders, acute viral diseases, lepromatous leprosy, B-CLL and inflammatory diseases
Process:
- Antibody specific for CD23 pre-coated onto microwells
- CD23 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody specific for CD23 is added to detect captured CD23 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | BC064417 |
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 2.2 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 2208 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human CD23,FCER2 ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction