missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 0.16 - 10 ng/mL |
| Assay Sensitivity | 0.09 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1380 |
| Gene Symbol | CR2 |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | C3b/C4b receptor, C3-binding protein, C3BR, C3DR, C4BR, CD35, CD35 antigen, complement component (3b/4b) receptor 1 (Knops blood group), complement component (3d/Epstein Barr virus) receptor 2, complement component 3d receptor 2, complement receptor 1, complement receptor type 1, complement receptor type 2, CR, CVID7, EBV receptor, KN, Knops blood group antigen, SLEB9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?