missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD21 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£173.00 - £464.00
Specifications
| Antigen | CD21 |
|---|---|
| Clone | 544411 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736335
![]() |
R&D Systems
MAB49091-100 |
100 μg |
£464.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18716035
![]() |
R&D Systems
MAB49091-SP |
25 μg |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for ImmunohistochemistrySpecifications
| CD21 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| C3DR, C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD21, Ile21-Arg971, Accession # P20023 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 544411 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P20023 | |
| 1380 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title