missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD200 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems BAM27242
This item is not returnable.
View return policy
Description
CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Specifications
| CD200/OX2 | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| P41217.3 | |
| CD200 | |
| Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 Accession # P41217.3 | |
| 250 μg | |
| Primary | |
| Detects human CD200 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CD200 is observed. | |
| Human | |
| Purified |
| ELISA | |
| 325523 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4345 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction