missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD19 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4867B-025
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD19 | |
| Monoclonal | |
| Biotin | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 930 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 4G7-2E3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| Human CLL cells | |
| 25 Tests | |
| Primary | |
| Detects human CD19 in flow cytometry. | |
| Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction