missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD164 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5790-SP
This item is not returnable.
View return policy
Description
CD164 Polyclonal specifically detects CD164 in Human samples. It is validated for Western Blot.
Specifications
| CD164 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD164, CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24, MGC-24v, MUC-24, Multi-glycosylated core protein 24, sialomucin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8763 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q04900 | |
| CD164 | |
| Mouse myeloma cell line NS0-derived recombinant human CD164 extracellular domain Asp24-Leu197 Accession # Q04900 | |
| 25 μg | |
| Primary | |
| Detects human CD164 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CD164 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction