missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD163 Quantikine ELISA Kit

Product Code. 18791934
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18791934 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18791934 Supplier R&D Systems™ Supplier No. DC1630

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CD163 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CD163 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD163 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56 - 100ng/mL
Assay Sensitivity 0.613ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Human CD163
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 9332
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.