Learn More
Human CD163 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 12 publications
Brand:  R&D Systems AF1607-SP
145.35 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18787067
  • £187.00 / 25µg
Description
CD163 Polyclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD163
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Goat
Affinity Purified
RUO
9332
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 1 ug/mL, Simple Western 10 ug/mL
CAA80543
CD163
Mouse myeloma cell line NS0-derived recombinant human CD163 (R&D Systems, Catalog # 1607-CD) Gly41-Ser1045 Accession # CAA80543
25 μg
Primary
Detects human CD163 in direct ELISAs and Western blots.