missing translation for 'onlineSavingsMsg'
Learn More

Human CD161 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  R&D Systems FAB7448N-100UG

Product Code. 18751888

  • £385.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD161
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q12918
KLRB1
BaF3 mouse pro-B cell line transfected with human CD161 Accession # Q12918
100 μg
Primary
Detects human CD161 in direct ELISAs.
Human
IgG2b
Flow Cytometry
702228
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3820
Reconstitution default not known
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)