missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD161 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7448T-100UG
This item is not returnable.
View return policy
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD161 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q12918 | |
| KLRB1 | |
| BaF3 mouse pro-B cell line transfected with human CD161 Accession # Q12918 | |
| RUO | |
| 3820 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 702228 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD161 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction