missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD160 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6177-SP
This item is not returnable.
View return policy
Description
CD160 Polyclonal specifically detects CD160 in Human samples. It is validated for Western Blot.
Specifications
| CD160 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BY55, BY55FLJ46513, CD160, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55, NK1, NK28 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 11126 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O95971 | |
| CD160 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD160 Ile27-Ser159 Accession # O95971 | |
| 25 μg | |
| Primary | |
| Detects human CD160 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse CD160 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction