missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD155/PVR Alexa Fluor™ 405-conjugated Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB11503V-100UG
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Flow Cytometry
Specifications
| CD155/PVR | |
| Monoclonal | |
| Alexa Fluor 405 | |
| P15151 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5817 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1072806 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
| Mouse myeloma cell line NS0-derived recombinant human CD155/PVR, Gly27-Asn343, Accession # P15151 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction