missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD151 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1884
This item is not returnable.
View return policy
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD151 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| BAA06229 | |
| CD151 | |
| NS0 mouse myeloma cell line transfected with human CD151 Met1-Tyr253 Accession # BAA06229 | |
| 500 μg | |
| Primary | |
| Detects human CD151. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 210127 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD151, CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, SFA-1, tetraspanin-24, tspan-24, TSPAN24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 977 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction