missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£235.00 - £411.00
Specifications
| Antigen | CD14 |
|---|---|
| Clone | 134620 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773099
![]() |
R&D Systems
FAB3832C-025 |
25 Tests |
£235.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707097
![]() |
R&D Systems
FAB3832C-100 |
100 Tests |
£411.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
| CD14 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| CD14 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD14 in direct ELISAs and Western blots. |
| 134620 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| Human | |
| P08571 | |
| 929 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title