missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 16 publications
£191.00 - £1785.00
Specifications
| Antigen | CD14 |
|---|---|
| Clone | 134620 |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 10 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773769
![]() |
R&D Systems
MAB3832-SP |
25 μg |
£191.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767519
![]() |
R&D Systems
MAB3832-100 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780917
![]() |
R&D Systems
MAB3832-500 |
500 μg |
£1785.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| CD14 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 10 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P08571 | |
| 929 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD14 in direct ELISAs and Western blots. |
| 134620 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| CD14 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title