missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD11b/Integrin alpha M Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£184.00 - £492.00
Specifications
| Antigen | CD11b |
|---|---|
| Clone | 238439 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724459
![]() |
R&D Systems
MAB16992-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18172831
![]() |
R&D Systems
MAB16992 |
500 μg |
£492.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Western Blot.Specifications
| CD11b | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| ITGAM | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 238439 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001139280 | |
| 3684 | |
| Mouse myeloma cell line NS0-derived recombinant human CD11b/Integrin alpha M Phe17-Asn1105 Accession # NP_001139280 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD11b/Integrin alpha M in direct ELISAs and Western blots. In Western blots no cross-reactivity with recombinant mouse (rm) Integrins alpha 2, alpha 3, alpha 4, alpha E or rhIntegrins alpha 5, alpha V, beta 1, beta 2, beta 3, or beta 6. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title