missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD11b/Integrin alpha M Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£191.00 - £352.00
Specifications
| Antigen | CD11b |
|---|---|
| Clone | ICRF44 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756888
![]() |
R&D Systems
FAB1699G-025 |
25 Tests |
£191.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18144551
![]() |
R&D Systems
FAB1699G-100 |
100 Tests |
£352.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Flow Cytometry.Specifications
| CD11b | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3684 | |
| Human rheumatoid synovial cells and monocytes | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| ICRF44 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| ITGAM | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Integrin alpha M/CD11b. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title