missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD117/c-kit Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF3267
This item is not returnable.
View return policy
Description
CD117/c-kit Polyclonal specifically detects CD117/c-kit in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| CD117/c-kit | |
| Polyclonal | |
| Unconjugated | |
| P10721 | |
| KIT | |
| Human CD117/c-kit synthetic peptide Asp716-Pro726 Accession # P10721 | |
| 50 μg | |
| Primary | |
| Detects human CD117/c-kit in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3815 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction