missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD112 (PVRL2) Fc Chimera, Recombinant Protein

Product Code. 17124173
Click to view available options
Quantity:
5 x 5 μg
5 x 50 μg
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 17124173

Brand: Invitrogen™ A42495

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HEK293 cells Recombinant Protein

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92692
For Use With (Application) Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot
Formulation protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 5819
Name Human CD112 (PVRL2) Fc Chimera
Quantity 5 x 5 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AI325026; AI987993; Cd112; Herpes virus entry mediator B; herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Mph; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; NECTIN2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
Gene Symbol NECTIN2
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.