missing translation for 'onlineSavingsMsg'
Learn More

Human CCRL2/CRAM-A/B Antibody, R&D Systems™
GREENER_CHOICE

Codice prodotto. 18183481
Cambia vista
Click to view available options
Quantity:
500 μg
Dimensione della confezione:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18183481 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18183481 Fornitore R&D Systems N. del fornitore MAB2350

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody has been used in 2 publications

CCRL2/CRAM-A/B Monoclonal specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifica

Antigen CCRL2/CRAM-A/B
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 152211
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001124382
Gene Alias ACKR5, C-C chemokine receptor-like 2, CCR11, CCR6, CCRL2, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CKRXMGC116710, CRAM, CRAMA, CRAM-A, CRAMB, CRAM-B, FLJ55815, HCR, HCRMGC34104, Putative MCP-1 chemokine receptor
Gene Symbols CCRL2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCRL2/CRAM-A/B Met1-Val356 (Val180Met) Accession # NP_001124382
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9034
Test Specificity Detects human CCRL2/CRAM-A/B. Stains human HCR-transfected cells but not irrelevant transfectants. It detects both the CRAM-A and CRAM-B isoforms.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.