missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£205.00 - £416.00
Specifications
| Antigen | CCR9 |
|---|---|
| Clone | 248621R |
| Dilution | Flow Cytometry 0.25μg/106 cells, Immunocytochemistry 8-25μg/mL, CyTOF-ready |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18198950
![]() |
R&D Systems
MAB1791R-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18119420
![]() |
R&D Systems
MAB1791R-100 |
100 μg |
£416.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| CCR9 | |
| Flow Cytometry 0.25μg/106 cells, Immunocytochemistry 8-25μg/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P51686 | |
| 10803 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 248621R | |
| CyTOF, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| Human CCR9 transfectants Met1-Leu369 Accession # P51686 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human CCR9 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title