missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR8 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1429-100
This item is not returnable.
View return policy
Description
CCR8 Monoclonal specifically detects CCR8 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.
Specifications
| CCR8 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 μg/10^6 cells, Neutralization 0.01-0.05 μg/mL, CyTOF-ready | |
| P51685 | |
| CCR8 | |
| BaF3 mouse pro-B cell line transfected with human CCR8 Met1-Leu355 Accession # P51685 | |
| 100 μg | |
| Primary | |
| Detects human CCR8. Neutralizes the bioactivity of human CCR8 and specifically stains human CCR8-transfected cell lines but not with the parent cell lines. | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 191704 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1237 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction