missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£191.00 - £460.00
Specifications
| Antigen | CCR6 |
|---|---|
| Clone | 53103R |
| Dilution | Flow Cytometry 0.25μg/106 cells, Immunohistochemistry 5-25μg/mL, CyTOF-ready |
| Applications | CyTOF, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18180191
![]() |
R&D Systems
MAB195R-SP |
25 μg |
£191.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18121441
![]() |
R&D Systems
MAB195R-100 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| CCR6 | |
| Flow Cytometry 0.25μg/106 cells, Immunohistochemistry 5-25μg/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P51684 | |
| 1235 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 53103R | |
| CyTOF, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
| NS0 mouse myeloma cell line transfected with human CCR6 Met1-Met374 Accession # P51684 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human CCR6. Stains human CCR6 transfectants but not the parental cell line in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title