missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1567R-100
This item is not returnable.
View return policy
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CCR4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Human CCR4 transfectants Met1-Leu360 Accession # P51679 | |
| Primary | |
| Detects human CCR4 in direct ELISAs | |
| Human | |
| Supernatant |
| CyTOF, Flow Cytometry | |
| 205410R | |
| Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| P51679 | |
| Mouse | |
| 100 μg | |
| 1233 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction