missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR10 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
£242.00 - £563.00
Specifications
| Antigen | CCR10/GPR2 |
|---|---|
| Clone | 314305 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743659
![]() |
R&D Systems
FAB3478A-025 |
25 Tests |
£242.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707716
![]() |
R&D Systems
FAB3478A-100 |
100 Tests |
£563.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR10/GPR2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
| CCR10 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detect human CCR10. Stains human CCR10-transfected cells but not irrelevant transfectants. |
| 314305 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| P46092 | |
| 2826 | |
| Y3 rat myeloid cell line transfected with human CCR10 Gln8-Asn362 Accession # P46092 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title