missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL7/MCP-3/MARC Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB282-SP
This item is not returnable.
View return policy
Description
CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL7/MCP-3/MARC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP-3, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6354 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, Flow Cytometry, CyTOF, ELISA | |
| 36320 | |
| Western Blot 1 ug/mL, Neutralization 1.5-4.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q7Z7Q8 | |
| CCL7 | |
| E. coli-derived recombinant human CCL7/MCP-3/MARC Gln34-109 Accession # Q7Z7Q8 | |
| 25 μg | |
| Primary | |
| Detects human CCL7/MCP-3/MARC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) CCL2/MCP-1, rhCCL8/MCP-2, or rhCCL13/MCP-4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction