Learn More
Human CCL7/MCP-3/MARC Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems IC282G-100UG
Product Code. 18723849
  • £408.00 / 100µg
Description
CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL7/MCP-3/MARC
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q7Z7Q8
CCL7
E. coli-derived recombinant human CCL7/MCP-3/MARC Gln34-109 Accession # Q7Z7Q8
RUO
6354
Human
IgG1
Flow Cytometry
36320
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP-3, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Mouse
100 μg
Primary
Detects human CCL7/MCP-3/MARC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) CCL2/MCP-1, rhCCL8/MCP-2, or rhCCL13/MCP-4 is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only (RUO)