missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCL5 (RANTES) Recombinant Protein

Product Code. p-7220360
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
This item is not returnable. View return policy

Product Code. 17676645

Brand: Invitrogen™ RP87853

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

RANTES is a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. It binds to CCR1, CCR3, CCR4 and CCR5 and is one of the major HIV suppressive factors produced by CD8+ T cells. Recombinant RANTES protein induces a dose dependent inhibition of different strains of HIV 1, HIV 2, and simian immunodeficiency virus (SIV). It is T cell and macrophage specific, induced by mitogens, and belongs to the intercrine beta (chemokine CC) family.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13501
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation 0.1% TFA with 26% acetonitrile and no preservative
Gene ID (Entrez) 6352
Molecular Weight (g/mol) 7.9 kD
Name Human CCL5 (RANTES) (aa 24-91)
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.1 EU/ μg cytokine
Gene Alias beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; eoCP; Eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; SCYA5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014)
Gene Symbol CCL5
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.