missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB507-100
This item is not returnable.
View return policy
Description
BMP-6 Monoclonal specifically detects BMP-6 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| BMP-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, Vgr-1, VGR1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 654 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization, ELISA | |
| 74219 | |
| Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q4VBA3 | |
| BMP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BMP-6 Arg296-His428 Accession # Q4VBA3 | |
| 100 μg | |
| Primary | |
| Detects human BMP-6 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BMP-2, rhBMP-4, rhBMP-5, or rhBMP-7 is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction