missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Human CCL3L1/LD78 beta ELISA Kit (Colorimetric) measures human CCL3L1/LD78 beta in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 78.13 - 5000 pg/mL |
| Assay Sensitivity | 46.88 pg/mL |
| Cross Reactivity | This kit recognizes Human CCL3L1 in samples. No significant cross-reactivity or interference between Human CCL3L1 and analogues was observed. |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 6349 |
| Gene Symbol | CCL3L1 |
| Host Species | Human |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?