missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL3/CCL4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2701-SP
This item is not returnable.
View return policy
Description
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| CCL3/CCL4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL3 | |
| E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147 | |
| 25 μg | |
| Primary | |
| Detects human CCL3/MIP-1 alpha in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed. Does not cross-react with rhCCL1, 2, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 2, 3, 5, 6, 7, CCL9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 93342 | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P10147 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6348 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction