missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL26/Eotaxin-3 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Specifications
Specifications
| Assay Range | 3.13 to 200 pg/mL |
| Assay Sensitivity | 0.215 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Heparin Plasma, Serum |
| Specificity | Natural and Recombinant Human Eotaxin-3 |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 10344 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4 A, MIP-4 alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1 Thymic stroma chemokine-1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction