missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL25 (TECK) Recombinant Protein, PeproTech®

Product Code. 17882553
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 17882553

Brand: Gibco™ 30045500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TECK is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15444
For Use With (Application) Control, Western Blot Control, ELISA Standard, Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 6370
Molecular Weight (g/mol) 14.2 kDa
Name Human CCL25 (TECK)
Purification Method Purified
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine
Common Name TECK
Gene Symbol CCL25
Biological Activity Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL
Content And Storage -20°C
Activity Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.