missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL25/TECK Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £492.00
Specifications
| Antigen | CCL25/TECK |
|---|---|
| Clone | 52529 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731029
![]() |
R&D Systems
MAB3341-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18171341
![]() |
R&D Systems
MAB3341 |
500 μg |
£492.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL25/TECK Monoclonal specifically detects CCL25/TECK in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| CCL25/TECK | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine | |
| CCL25 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 52529 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAB69981 | |
| 6370 | |
| E. coli-derived recombinant human CCL25/TECK Gln24-Leu150 Accession # AAB69981 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCL25/TECK in direct ELISAs and Western blots. Shows less than 1% cross-reactivity with recombinant mouse CCL25 and recombinant rat CINC-1 and no cross-reactivity with recombinant human (rh)CCL28, recombinant mouse (rm)CCL28, rhMPIF-1, rh6Ckine, rm6Ckine, rhEotaxin, or rmEotaxin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title