missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL24/Eotaxin-2/MPIF-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB343-SP
This item is not returnable.
View return policy
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2, MPIF-2, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6369 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 61016 | |
| Western Blot 1 ug/mL, Neutralization 0.2-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAB51135 | |
| CCL24 | |
| E. coli-derived recombinant human CCL24/Eotaxin-2/MPIF-2 Val27-Cys119 Accession # AAB51135 | |
| 25 μg | |
| Primary | |
| Detects human CCL24/Eotaxin-2/MPIF-2 in ELISAs and Western blots. In sandwich ELISAs, less than 0.04% cross-reactivity with recombinant human (rh) Eotaxin, rhMIP-1 alpha , and rhMCP-3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction