missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL23/Ck beta 8-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB508-SP
This item is not returnable.
View return policy
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL23/Ck beta 8-1/MIP3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C6 beta-chemokine, chemokine (C-C motif) ligand 23, Ck beta 8-1, CKb8, CKB-8, Ckb81, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6368 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 103952 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAD10846 | |
| CCL23 | |
| E. coli-derived recombinant human CCL23/CK beta 8-1 Met46-Asn137 Accession # AAD10846 | |
| 25 μg | |
| Primary | |
| Detects human CCL23/Ck beta 8-1 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with human CCL23/CK beta 8 isoform is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction