missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CCL20 (P78556, 27 a.a. - 96 a.a.) Partial Recombinant Protein
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Sequence: ASNFDCCLGYTDRILHPKFIVGFTRQLANEGCDINAIIFHTKKKLSVCANPKQTWVKYIVRLLSKKVKNM
Specifications
Specifications
| Accession Number | P78556 |
| For Use With (Application) | Functional Study, SDS-PAGE |
| Formulation | Lyophilized |
| Gene ID (Entrez) | 6364 |
| Molecular Weight (g/mol) | 8kDa |
| Name | CCL20 (Human) Recombinant Protein |
| Preparation Method | Escherichia coli expression system |
| Purification Method | Ion exchange column and HPLC reverse phase column |
| Quantity | 25 μg |
| Immunogen | ASNFDCCLGYTDRILHPKFIVGFTRQLANEGCDINAIIFHTKKKLSVCANPKQTWVKYIVRLLSKKVKNM |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction