missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCL2 (MCP-1) Recombinant Protein

Product Code. 11589161
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11589161 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11589161 Supplier Invitrogen™ Supplier No. RP8648

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

CCL2 (human) Recombinant Protein, RP-8648, from Invitrogen.

Monocyte Chemotactic Protein 1 (MCP-1), also known as CCL2, is thought to be produced by injured or infected tissues. MCP-1 signals through G protein-coupled receptors, CCR2 and CCR4, to recruit memory T cells, monocytes and dendritic cells. Recombinant human MCP-1 is a non-glycosylated protein containing 76 amino acids, with a molecular weight of 8.6 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human monocytes or THP-1 cells at concentrations of 4-15 ng/mL. The specific activity of this protein is 2.5x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13500
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 6347
Molecular Weight (g/mol) 8.6
Name Human CCL2 (MCP-1)
Quantity 20 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-MCP-1
Common Name Human CCL2 (MCP-1)
Gene Symbol CCL2
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.