missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCL2 (MCP-1) Recombinant Protein

Product Code. 11508991
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508991 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11508991 Supplier Invitrogen™ Supplier No. RP75664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human CCL2 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13500
Concentration 0.225 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 6347
Name Human CCL2 (MCP-1)
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1 A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1 A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Common Name CCL2 (MCP-1)
Gene Symbol CCL2
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Gln24-Thr99 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.