missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL2 (MCP-1) Recombinant Protein

Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC1014

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10680354

  • £92.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human CCL2 (MCP-1) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

MCP-1 is structurally related to the CC subfamily of cytokines. Members of this subfamily are characterized by two adjacent cysteines. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
8 kDa
10μg
Human
Carrier-Free
10 μg
E. coli
Recombinant, Ligand
MCP⁄MCAF Chemokines
Gibco™
6347
Sequential Chromatography
MCP-1 (MCAF)
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Chemokines & Receptors
Lyophilized
Signal Transduction, Inflammation, Bone Research
98 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.