missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL19/MIP-3 beta QuicKit ELISA
Specifications
Specifications
| Assay Range | 7.81 tp 500 pg/mL |
| Assay Sensitivity | 0.695 pg/mL |
| Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA), Plasma (Heparin) |
| Specificity | Natural and recombinant human MIP-3β |
| Target Species | Human |
| Gene ID (Entrez) | 6363 |
| Quantity | 1 Pack (6 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELC, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP3 beta, MIP-3 beta, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?