missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL18/PARC Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems BAF394
This item is not returnable.
View return policy
Description
CCL18/PARC Polyclonal specifically detects CCL18/PARC in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, ELISA Detection (Matched Antibody Pair).
Specifications
| CCL18/PARC | |
| Polyclonal | |
| Biotin | |
| P55774 | |
| CCL18 | |
| E. coli-derived recombinant human CCL18/PARC Ala21-Ala89 Accession # P55774 | |
| 50 μg | |
| Primary | |
| Detects human CCL18/PARC in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant human (rh) HCC-1, rhMPIF-1, rhMIP-1 alpha , recombinant mouse (rm) MIP-1 alpha , rhMIP-1 beta , rmMIP-1 beta , recombinant viral (rv) MIP-I, rvMIP-II, and rhMIP-1 delta is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| Alternative macrophage activation-associated CC chemokine 1, AMAC-1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4, MIP-4C-C motif chemokine 18, PARC, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6362 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction